Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012157.3 | NP_036289.3 | 7 | 492 | ||
XM_005265014.2 | XP_005265071.1 | 7 | 512 | |||
XM_005265015.3 | XP_005265072.1 | 7 | 732 | |||
XM_005265016.2 | XP_005265073.1 | 7 | 476 | |||
XM_005265017.3 | XP_005265074.1 | 7 | 508 | |||
XM_011533559.2 | XP_011531861.1 | 8 | 677 | |||
XM_017006092.1 | XP_016861581.1 | 8 | 569 | |||
XM_017006093.1 | XP_016861582.1 | 7 | 559 | |||
XM_017006094.1 | XP_016861583.1 | 8 | 592 | |||
GenBank mRNA | AF174589.1 | - | 8 | 463 | ||
AF176518.1 | - | 7 | 427 | |||
AF186273.1 | - | 7 | 442 | |||
AK001271.1 | - | 7 | 492 | |||
AK001438.1 | - | 7 | 460 | |||
AK294209.1 | - | 4 | 243 | |||
AK297534.1 | - | 7 | 476 | |||
AK299665.1 | - | 8 | 575 | |||
AK302218.1 | - | 9 | 720 | |||
AK303764.1 | - | 5 | 352 | |||
AK316229.1 | - | 7 | 513 | |||
AK316383.1 | - | 8 | 632 | |||
AU124816.1 | - | 7 | 492 | |||
BC031556.1 | - | 7 | 436 | 5197151 | ||
CR457121.1 | - | 7 | 421 |