Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012162.3 | NP_036294.2 | 7 | 1239 | ||
NM_024555.5 | NP_078831.4 | 7 | 1221 | |||
GenBank mRNA | AF370422.1 | - | 6 | 2552 | ||
AK026541.1 | - | 7 | 1170 | |||
AK223088.1 | - | 7 | 1170 | |||
AL713783.1 | - | 3 | 1450 | |||
BC033066.1 | - | 7 | 1228 | 5516945 |