Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004055.1 | NP_001004055.1 | 6 | 922 | ||
NM_012163.2 | NP_036295.1 | 6 | 1087 | |||
XM_017023126.1 | XP_016878615.1 | 6 | 1351 | |||
XM_017023127.1 | XP_016878616.1 | 6 | 1602 | |||
XM_017023128.1 | XP_016878617.1 | 6 | 1407 | |||
XM_017023129.1 | XP_016878618.1 | 6 | 2493 | |||
XM_017023130.1 | XP_016878619.1 | 6 | 1075 | |||
XM_017023131.1 | XP_016878620.1 | 6 | 934 | |||
XM_017023132.1 | XP_016878621.1 | 6 | 1237 | |||
XM_017023133.1 | XP_016878622.1 | 6 | 1263 | |||
XM_017023134.1 | XP_016878623.1 | 6 | 1042 | |||
GenBank mRNA | AF176701.1 | - | 6 | 932 | ||
AK096101.1 | - | 4 | 1559 | |||
AK316602.1 | - | 6 | 935 | |||
BC018785.1 | - | 6 | 1087 | 4875646 |