Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012175.3 | NP_036307.2 | 2 | 171 | ||
NM_033406.2 | NP_208385.1 | 2 | 171 | |||
XM_011519980.1 | XP_011518282.1 | 2 | 177 | |||
XM_011519981.2 | XP_011518283.1 | 2 | 177 | |||
XM_011519982.1 | XP_011518284.1 | 2 | 150 | |||
XM_017017533.1 | XP_016873022.1 | 2 | 150 | |||
GenBank mRNA | AB209039.1 | - | 1 | 3556 | ||
AF174595.1 | - | 2 | 131 | |||
AF176702.1 | - | 2 | 128 | |||
AK001943.1 | - | 2 | 162 | |||
AK128454.1 | - | 2 | 149 | |||
AK304866.1 | - | 2 | 166 | |||
AL136625.1 | - | 2 | 154 | |||
AL832826.1 | - | 1 | 843 | |||
BC039291.1 | - | 2 | 171 | |||
BC046110.1 | - | 2 | 157 | 5549548 | ||
BG716664.1 | - | 2 | 174 |