Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012198.3 | NP_036330.1 | 2 | 332 | ||
XM_005246446.2 | XP_005246503.1 | 2 | 273 | |||
XM_006712398.3 | XP_006712461.1 | 2 | 505 | |||
XM_006712400.3 | XP_006712463.1 | 3 | 583 | |||
XM_006712401.3 | XP_006712464.1 | 2 | 584 | |||
XM_011510926.2 | XP_011509228.1 | 2 | 284 | |||
XM_011510927.2 | XP_011509229.1 | 2 | 279 | |||
XM_017003764.1 | XP_016859253.1 | 3 | 596 | |||
XM_017003765.1 | XP_016859254.1 | 3 | 682 | |||
XM_017003766.1 | XP_016859255.1 | 3 | 479 | |||
XM_017003767.1 | XP_016859256.1 | 2 | 355 | |||
XM_017003768.1 | XP_016859257.1 | 2 | 350 | |||
XM_017003769.1 | XP_016859258.1 | 2 | 505 | |||
GenBank mRNA | AB209768.1 | - | 2 | 221 | ||
AK295413.1 | - | 2 | 265 | |||
AK312349.1 | - | 2 | 281 | |||
BC005214.1 | - | 2 | 255 | 3996889 | ||
M81637.1 | - | 2 | 275 |