Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303277.1 | NP_001290206.1 | 6, 7 | 552 | ||
NM_012278.2 | NP_036410.1 | 9, 10 | 805 | |||
XM_017029405.1 | XP_016884894.1 | 7, 8 | 1075 | |||
GenBank mRNA | AF110225.1 | - | 9, 10 | 759 | ||
AF140690.1 | - | 9, 10 | 775 | |||
BC108901.2 | - | 8, 9 | 1032 | 40004692 | ||
BP372847.1 | - | 6, 7 | 550 |