Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303277.1 | NP_001290206.1 | 8 | 864 | ||
NM_012278.2 | NP_036410.1 | 11 | 1117 | |||
XM_017029405.1 | XP_016884894.1 | 9 | 1387 | |||
GenBank mRNA | AF110225.1 | - | 11 | 1071 | ||
AF140690.1 | - | 11 | 1087 | |||
BC108901.2 | - | 10 | 1344 | 40004692 |