Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282157.1 | NP_001269086.1 | 9 | 1313 | ||
NM_012280.3 | NP_036412.1 | 13 | 1735 | |||
NM_177439.2 | NP_803188.1 | 14 | 1835 | |||
XM_005272595.1 | XP_005272652.1 | 13 | 1684 | |||
GenBank mRNA | AF063015.1 | - | 12 | 1322 | ||
AJ005892.1 | - | 13 | 1572 | |||
AK024023.1 | - | 14 | 1695 | |||
AK297486.1 | - | 9 | 1137 | |||
BC021047.1 | - | 13 | 1503 | |||
BC023584.1 | - | 13 | 1501 | 3619796 |