Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004423.3 | NP_004414.3 | 15 | 4326 | ||
XM_005247172.2 | XP_005247229.1 | 16 | 3674 | |||
XM_011512513.2 | XP_011510815.1 | 12 | 4114 | |||
GenBank mRNA | D86963.1 | - | 15 | 4326 |