Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012316.4 | NP_036448.1 | 14 | 1899 | ||
XM_005270711.3 | XP_005270768.1 | 14 | 2260 | |||
GenBank mRNA | AK002111.1 | - | 14 | 1886 | ||
AK128609.1 | - | 14 | 1862 | |||
AK295615.1 | - | 14 | 2014 | |||
AK301714.1 | - | 14 | 1853 | |||
BC020520.1 | - | 14 | 1874 | 3913864 |