Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012317.3 | NP_036449.1 | 1 | 1080 | ||
GenBank mRNA | AB019527.1 | - | 1 | 991 | ||
BC003104.1 | - | 1 | 965 | 3503444 | ||
HQ343285.1 | - | 2 | 507 |