Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099406.1 | NP_001092876.1 | 2 | 388 | ||
NM_012319.3 | NP_036451.3 | 3 | 1186 | |||
XM_011525900.1 | XP_011524202.1 | 3 | 1619 | |||
XM_011525901.1 | XP_011524203.1 | 4 | 2303 | |||
GenBank mRNA | AK299102.1 | - | 2 | 219 | ||
AK301539.1 | - | 2 | 338 | |||
AK304079.1 | - | 3 | 1110 | |||
BC039498.1 | - | 2 | 379 | 5552785 | ||
BX336036.2 | - | 2 | 157 | |||
U41060.2 | - | 4 | 1015 |