Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012344.3 | NP_036476.1 | 4 | 1263 | ||
XM_005246156.3 | XP_005246213.1 | 3 | 992 | |||
XM_006711876.3 | XP_006711939.1 | 4 | 1208 | |||
XM_006711877.3 | XP_006711940.1 | 4 | 1199 | |||
XM_017003738.1 | XP_016859227.1 | 4 | 1259 | |||
GenBank mRNA | AY429107.1 | - | 4 | 1189 | ||
BC022501.1 | - | 4 | 1224 | 4792730 | ||
BC037776.2 | - | 5 | 1177 | 4817029 | ||
Y10148.1 | - | 4 | 1225 |
Set Membership: |
G-protein coupled receptor |