Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012345.2 | NP_036477.2 | 1 | 374 | ||
GenBank mRNA | AF159548.1 | - | 1 | 332 | ||
AK314242.1 | - | 1 | 367 | |||
BC001436.1 | - | 1 | 356 | |||
DA701559.1 | - | 1 | 375 |