Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 3 | 394 | ||
NM_012388.3 | NP_036520.1 | 3 | 560 | |||
NR_132350.1 | 4 | 568 | ||||
NR_132351.1 | 2, 3 | 556 | ||||
NR_132353.1 | 2 | 418 | ||||
NR_132354.1 | 1, 2 | 414 | ||||
NR_132356.1 | 3 | 262 | ||||
NR_132357.1 | 3 | 262 | ||||
GenBank mRNA | AF080470.1 | - | 3 | 464 | ||
AK057545.1 | - | 2, 3 | 300 | |||
AK128626.1 | - | 4 | 537 | |||
AK315581.1 | - | 3 | 278 | |||
AL596952.1 | - | 3 | 333 | |||
BC004819.1 | - | 3 | 281 | 3450397 | ||
DA798595.1 | - | 1, 2 | 186 | |||
DA859617.1 | - | 3 | 210 | |||
DB212570.1 | - | 2 | 305 | |||
DB289156.1 | - | 3 | 205 |