Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA2 SIRNA Assays ›
Gene Symbol:
PPFIA2
Gene Name:
PTPRF interacting protein alpha 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 81257975 - 81759748 on Build GRCh38
UniGene ID:
Hs.506216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13393
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220473.2
NP_001207402.1
2
498
NM_001220474.2
NP_001207403.1
2
498
NM_001220475.2
NP_001207404.1
2
310
NM_001220476.2
NP_001207405.1
2
310
NM_003625.4
NP_003616.2
3
361
XM_011538906.2
XP_011537208.1
2
492
XM_011538907.2
XP_011537209.1
2
492
XM_017020079.1
XP_016875568.1
3
345
XM_017020080.1
XP_016875569.1
2
492
XM_017020081.1
XP_016875570.1
2
492
XM_017020082.1
XP_016875571.1
3
345
XM_017020083.1
XP_016875572.1
2
492
XM_017020084.1
XP_016875573.1
2
492
XM_017020085.1
XP_016875574.1
2
492
XM_017020086.1
XP_016875575.1
3
345
XM_017020087.1
XP_016875576.1
2
479
XM_017020088.1
XP_016875577.1
3
779
XM_017020089.1
XP_016875578.1
2
492
XM_017020090.1
XP_016875579.1
3
345
XM_017020091.1
XP_016875580.1
2
492
XM_017020092.1
XP_016875581.1
2
492
XM_017020093.1
XP_016875582.1
2
492
XM_017020094.1
XP_016875583.1
3
345
XM_017020095.1
XP_016875584.1
2
492
XM_017020096.1
XP_016875585.1
2
492
XM_017020097.1
XP_016875586.1
3
345
XM_017020098.1
XP_016875587.1
3
345
XM_017020099.1
XP_016875588.1
2
492
XM_017020100.1
XP_016875589.1
2
492
XM_017020101.1
XP_016875590.1
3
345
XM_017020102.1
XP_016875591.1
2
479
XM_017020103.1
XP_016875592.1
2
492
XM_017020104.1
XP_016875593.1
2
492
XM_017020105.1
XP_016875594.1
2
492
XM_017020106.1
XP_016875595.1
2
492
XM_017020107.1
XP_016875596.1
3
345
XM_017020108.1
XP_016875597.1
3
345
XM_017020109.1
XP_016875598.1
2
492
XM_017020110.1
XP_016875599.1
3
779
XM_017020111.1
XP_016875600.1
2
492
GenBank mRNA
AB210009.1
-
2
496
AF034799.1
-
3
369
AK126971.1
-
2
458
AK307524.1
-
3
345
BC104912.1
-
2
294
8143915
BC143485.1
-
2
294
9051993
DA499362.1
-
2
499
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
neurotransmitter secretion
glutamate secretion
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-wjdsd:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline