Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136138.2 | NP_001129610.1 | 5 | 837 | ||
NM_001287218.1 | NP_001274147.1 | 7 | 951 | |||
NM_001287219.1 | NP_001274148.1 | 7 | 990 | |||
NM_001287220.1 | NP_001274149.1 | 6 | 873 | |||
NM_001287221.1 | NP_001274150.1 | 5 | 752 | |||
NM_012424.4 | NP_036556.2 | 6 | 873 | |||
XM_005273095.1 | XP_005273152.1 | 6 | 860 | |||
XM_005273096.1 | XP_005273153.1 | 7 | 971 | |||
XM_017001024.1 | XP_016856513.1 | 4 | 534 | |||
XM_017001025.1 | XP_016856514.1 | 7 | 924 | |||
XM_017001026.1 | XP_016856515.1 | 7 | 917 | |||
XM_017001027.1 | XP_016856516.1 | 8 | 1049 | |||
XM_017001028.1 | XP_016856517.1 | 8 | 1049 | |||
XM_017001030.1 | XP_016856519.1 | 7 | 971 | |||
XM_017001031.1 | XP_016856520.1 | 8 | 1049 | |||
XM_017001032.1 | XP_016856521.1 | 7 | 926 | |||
XM_017001033.1 | XP_016856522.1 | 7 | 971 | |||
XM_017001034.1 | XP_016856523.1 | 8 | 1049 | |||
GenBank mRNA | AB070706.1 | - | 6 | 701 | ||
AF477978.1 | - | 6 | 850 | |||
AK122921.1 | - | 5 | 808 | |||
AK122989.1 | - | 6 | 860 | |||
AK299298.1 | - | 7 | 976 | |||
BC104769.1 | - | 6 | 787 | 8143772 | ||
BC143473.1 | - | 6 | 787 | |||
BX647303.1 | - | 4 | 515 |