Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114172.1 | NP_001107644.1 | 3 | 448 | ||
NM_001303427.1 | NP_001290356.1 | 2 | 353 | |||
NM_001303428.1 | NP_001290357.1 | 3 | 292 | |||
NM_001303429.1 | NP_001290358.1 | 2 | 947 | |||
NM_003629.3 | NP_003620.3 | 2 | 947 | |||
XM_017002614.1 | XP_016858103.1 | 3 | 325 | |||
GenBank mRNA | AF028785.1 | - | 2 | 882 | ||
AK302049.1 | - | 2 | 346 | |||
AK313726.1 | - | 3 | 460 | |||
BC005381.1 | - | 2 | 418 | |||
BC021622.1 | - | 2 | 881 | 3910221 | ||
BX537388.1 | - | 3 | 292 | |||
CR749649.1 | - | 2 | 868 | |||
D88532.1 | - | 2 | 611 | |||
DB047431.1 | - | 2 | 353 |