Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012444.2 | NP_036576.1 | 5 | 539 | ||
NM_198265.1 | NP_937998.1 | 4 | 425 | |||
XM_005260379.3 | XP_005260436.1 | 5 | 557 | |||
XM_005260380.3 | XP_005260437.1 | 5 | 557 | |||
XM_005260381.3 | XP_005260438.1 | 5 | 557 | |||
XM_005260382.4 | XP_005260439.1 | 5 | 557 | |||
XM_011528756.2 | XP_011527058.1 | 4 | 443 | |||
XM_011528757.2 | XP_011527059.1 | 5 | 391 | |||
GenBank mRNA | AF149310.1 | - | 5 | 479 | ||
AF169385.1 | - | 5 | 554 | |||
BC033591.1 | - | 4 | 387 | 5167214 |