Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012459.2 | NP_036591.2 | 2 | 435 | ||
NR_028383.1 | 3 | 768 | ||||
GenBank mRNA | AF150087.1 | - | 2 | 372 | ||
AF165967.1 | - | 3 | 678 | |||
AK025397.1 | - | 2 | 393 | |||
BC000711.2 | - | 3 | 715 | 2823930 | ||
BC105986.1 | - | 2 | 403 | 4510312 | ||
BC106067.1 | - | 2 | 403 | 4708781 |