Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013238.2 | NP_037370.2 | 5 | 752 | ||
GenBank mRNA | AF126743.1 | - | 6 | 774 | ||
AJ270691.1 | - | 2 | 118 | |||
AK311866.1 | - | 5 | 388 | |||
AY248898.1 | - | 5 | 752 | |||
AY512565.1 | - | 5 | 445 | |||
BC010910.1 | - | 5 | 481 | 4332619 | ||
BC095400.1 | - | 5 | 371 | 30530999 |