Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318202.1 | NP_001305131.1 | 9, 10 | 1098 | ||
NM_013241.2 | NP_037373.2 | 9, 10 | 1098 | |||
XM_011523043.2 | XP_011521345.1 | 9, 10 | 1314 | |||
XM_011523044.1 | XP_011521346.1 | 9, 10 | 1043 | |||
XM_011523045.2 | XP_011521347.1 | 9, 10 | 808 | |||
GenBank mRNA | AB041046.1 | - | 9, 10 | 1068 | ||
AB209584.1 | - | 7, 8 | 1567 | |||
AF113615.1 | - | 9, 10 | 1004 | |||
AK304232.1 | - | 9, 10 | 1041 | |||
AY192154.1 | - | 9, 10 | 986 | |||
BC033084.1 | - | 9, 10 | 1043 | 5456031 | ||
KJ658417.1 | - | 9, 10 | 986 |