Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318202.1 | NP_001305131.1 | 15 | 1827 | ||
NM_013241.2 | NP_037373.2 | 13 | 1749 | |||
XM_006721180.1 | XP_006721243.1 | 4 | 402 | |||
XM_011523043.2 | XP_011521345.1 | 15 | 2043 | |||
XM_011523044.1 | XP_011521346.1 | 15 | 1772 | |||
XM_011523045.2 | XP_011521347.1 | 15 | 1537 | |||
GenBank mRNA | AB041046.1 | - | 13 | 1719 | ||
AB209584.1 | - | 11 | 2218 | |||
AF113615.1 | - | 13 | 1655 | |||
AK301157.1 | - | 3 | 460 | |||
AK304232.1 | - | 13 | 1684 | |||
AY192154.1 | - | 13 | 1637 | |||
BC033084.1 | - | 13 | 1694 | 5456031 | ||
KJ658417.1 | - | 15 | 1715 |