Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165257.1 | NP_001158729.1 | 11 | 1744 | ||
NM_013243.3 | NP_037375.2 | 12 | 1797 | |||
GenBank mRNA | AF078851.1 | - | 12 | 1679 | ||
AF453583.1 | - | 12 | 1802 | |||
AK075314.1 | - | 12 | 1656 | |||
AK290175.1 | - | 12 | 1633 | |||
AK296466.1 | - | 11 | 1583 | |||
AY359093.1 | - | 12 | 1658 | |||
BC009511.2 | - | 6 | 689 | |||
BC014539.1 | - | 12 | 1433 | 4283082 |