Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301167.1 | NP_001288096.1 | 6 | 1743 | ||
NM_013252.2 | NP_037384.1 | 7 | 1788 | |||
XM_011515995.2 | XP_011514297.1 | 5 | 1675 | |||
XM_017011915.1 | XP_016867404.1 | 6 | 1746 | |||
XM_017011917.1 | XP_016867406.1 | 6 | 1741 | |||
GenBank mRNA | AJ271684.1 | - | 7 | 1788 |