Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018057.1 | NP_001018067.1 | 2 | 358 | ||
NM_013253.4 | NP_037385.2 | 3 | 526 | |||
NM_015881.5 | NP_056965.3 | 3 | 540 | |||
XM_006718178.2 | XP_006718241.1 | 3 | 353 | |||
XM_017017553.1 | XP_016873042.1 | 3 | 487 | |||
XM_017017554.1 | XP_016873043.1 | 3 | 505 | |||
XM_017017555.1 | XP_016873044.1 | 3 | 353 | |||
GenBank mRNA | AB033421.1 | - | 2 | 319 | ||
AB034203.1 | - | 3 | 498 | |||
AB057591.1 | - | 3 | 526 | |||
AF177396.1 | - | 2 | 338 | |||
AK090952.1 | - | 445 | ||||
AK092979.1 | - | 3 | 317 | |||
AK289897.1 | - | 3 | 367 | |||
AK295441.1 | - | 2 | 302 | |||
AK295539.1 | - | 3 | 461 | |||
AL535720.3 | - | 2 | 357 | |||
AY358378.1 | - | 3 | 453 | |||
AY587550.1 | - | 2 | 301 | |||
BC007660.2 | - | 3 | 539 | 3347453 | ||
CB156451.1 | - | 3 | 513 |