Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018057.1 | NP_001018067.1 | 7 | 1257 | ||
NM_013253.4 | NP_037385.2 | 8 | 1425 | |||
NM_015881.5 | NP_056965.3 | 8 | 1439 | |||
XM_006718178.2 | XP_006718241.1 | 8 | 1252 | |||
XM_017017553.1 | XP_016873042.1 | 8 | 1428 | |||
XM_017017554.1 | XP_016873043.1 | 8 | 1446 | |||
XM_017017555.1 | XP_016873044.1 | 8 | 1294 | |||
GenBank mRNA | AB033421.1 | - | 7 | 1217 | ||
AB034203.1 | - | 8 | 1397 | |||
AB057591.1 | - | 8 | 1425 | |||
AF052161.1 | - | 1 | 123 | |||
AF177396.1 | - | 7 | 1237 | |||
AK090952.1 | - | 979 | ||||
AK098756.1 | - | 2 | 415 | |||
AK225156.1 | - | 7 | 1209 | |||
AY358378.1 | - | 8 | 1352 | |||
BC007660.2 | - | 8 | 1438 | 3347453 | ||
BQ690088.1 | - | 2 | 504 | |||
BQ690888.1 | - | 2 | 504 |