Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013254.3 | NP_037386.1 | 20, 21 | 2305 | ||
XM_005268809.1 | XP_005268866.1 | 20, 21 | 2383 | |||
XM_005268810.1 | XP_005268867.1 | 20, 21 | 2365 | |||
GenBank mRNA | AF174536.1 | - | 19, 20 | 2146 | ||
AF191838.1 | - | 20, 21 | 2208 | |||
AK002192.1 | - | 12, 13 | 1131 | |||
AK291039.1 | - | 20, 21 | 2250 | |||
AK303683.1 | - | 15, 16 | 1522 | |||
BC034950.2 | - | 20, 21 | 2245 | 4824988 |