Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033578.2 | NP_001028750.1 | 11 | 1325 | ||
NM_013257.4 | NP_037389.4 | 11 | 1048 | |||
NM_170709.2 | NP_733827.2 | 11 | 1048 | |||
GenBank mRNA | AF085233.2 | - | 10 | 839 | ||
AF169035.1 | - | 10 | 809 | |||
AK074666.1 | - | 11 | 1002 | |||
AK223523.1 | - | 11 | 1017 | |||
AK291428.1 | - | 11 | 1325 | |||
BC015326.1 | - | 10 | 842 | 4391699 | ||
CR536581.1 | - | 8 | 587 | |||
CR933673.1 | - | 11 | 982 |
Gene Symbol: C8orf44-SGK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204173.1 | NP_001191102.1 | 13 | 1419 |