Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPSNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202502.1 | NP_001189431.1 | 2 | 160 | ||
NM_003634.3 | NP_003625.2 | 2 | 461 | |||
GenBank mRNA | AJ001258.1 | - | 2 | 460 | ||
AK098367.1 | - | 2 | 215 | |||
AK290953.1 | - | 2 | 214 | |||
AK298816.1 | - | 2 | 162 | |||
AK314407.1 | - | 2 | 232 | |||
BC002371.2 | - | 2 | 211 | 2822369 | ||
BC006473.1 | - | 2 | 211 | 2822369 | ||
BC018869.2 | - | 1 | 103 | |||
DA133702.1 | - | 2 | 279 |