Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013264.4 | NP_037396.3 | 12 | 1696 | ||
XM_011542790.2 | XP_011541092.1 | 12 | 1600 | |||
XM_011542791.2 | XP_011541093.1 | 12 | 1648 | |||
XM_017017626.1 | XP_016873115.1 | 12 | 1637 | |||
GenBank mRNA | AF155140.2 | - | 12 | 1590 | ||
BC035388.1 | - | 12 | 1542 | 5166189 | ||
BC050360.1 | - | 12 | 1566 | 5742404 | ||
CD657404.1 | - | 4 | 641 |