Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013276.2 | NP_037408.2 | 7 | 2799 | ||
GenBank mRNA | AF163573.1 | - | 7 | 2799 | ||
AK055609.1 | - | 7 | 2721 | |||
AK057020.1 | - | 1 | 1023 | |||
AK057040.1 | - | 1 | 1646 | |||
AL832420.1 | - | 7 | 2734 | |||
BC020543.1 | - | 7 | 2751 | 4413491 | ||
BC111366.1 | - | 1 | 677 |
Gene Symbol: TRPV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF308801.1 | - | 485 |