Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003635.3 | NP_003626.1 | 3, 4 | 1802 | ||
XM_005270255.3 | XP_005270312.1 | 3, 4 | 1802 | |||
XM_011540310.2 | XP_011538612.1 | 2, 3 | 2371 | |||
XM_017016857.1 | XP_016872346.1 | 3, 4 | 1802 | |||
GenBank mRNA | AB208870.1 | - | 1, 2 | 959 | ||
AK298957.1 | - | 1, 2 | 101 | |||
AK303648.1 | - | 4, 5 | 1600 | |||
BC035711.1 | - | 3, 4 | 1555 | 5587622 | ||
BC110588.1 | - | 1, 2 | 1034 | 40011898 | ||
BC110589.1 | - | 1, 2 | 1034 | 40012703 | ||
U36601.1 | - | 1, 2 | 1025 |