Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013306.4 | NP_037438.2 | 3, 4 | 387 | ||
NM_147777.3 | NP_680086.2 | 3, 4 | 387 | |||
GenBank mRNA | AB209439.1 | - | 3, 4 | 255 | ||
AF001435.1 | - | 3, 4 | 313 | |||
AF175267.2 | - | 3, 4 | 296 | |||
AF175268.1 | - | 3, 4 | 257 | |||
AK315034.1 | - | 3, 4 | 318 | |||
AK315642.1 | - | 3, 4 | 328 | |||
BC009897.2 | - | 3, 4 | 305 | 3510379 | ||
BC012767.2 | - | 3, 4 | 357 | 3678551 | ||
BC014520.2 | - | 3, 4 | 357 | 3678576 | ||
BT006631.1 | - | 3, 4 | 257 | |||
DA079695.1 | - | 3, 4 | 388 |
Gene Symbol: ARL2-SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037650.1 | 6, 7 | 911 | |||
GenBank mRNA | AK096447.1 | - | 6, 7 | 845 |