Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013313.4 | NP_037445.1 | 5 | 1601 | ||
NR_130910.1 | 5 | 1985 | ||||
GenBank mRNA | AB098734.1 | - | 5 | 1598 | ||
BC034486.1 | - | 5 | 1460 | 4826997 | ||
BQ422551.1 | - | 308 | ||||
CR933630.1 | - | 4 | 5705 |