Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013323.2 | NP_037455.2 | 7 | 1332 | ||
NM_152244.1 | NP_689450.1 | 8 | 1419 | |||
XM_005257260.3 | XP_005257317.1 | 7 | 1099 | |||
XM_005257261.3 | XP_005257318.1 | 7 | 1366 | |||
XM_005257262.3 | XP_005257319.1 | 7 | 1160 | |||
XM_011524697.2 | XP_011522999.1 | 8 | 1202 | |||
GenBank mRNA | AF121861.1 | - | 7 | 1352 | ||
AK023932.1 | - | 8 | 1419 | |||
AK091852.1 | - | 8 | 1317 | |||
AK296095.1 | - | 5 | 1226 | |||
AK296569.1 | - | 7 | 1300 | |||
AK298551.1 | - | 7 | 1155 | |||
AK316374.1 | - | 7 | 1152 | |||
BC000768.2 | - | 7 | 1149 | 2963905 | ||
BC103721.1 | - | 7 | 1138 | 6162638 |