Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013351.1 | NP_037483.1 | 6 | 2140 | ||
XM_011524698.2 | XP_011523000.1 | 7 | 2207 | |||
GenBank mRNA | AF093098.1 | - | 6 | 2140 | ||
BC039739.2 | - | 6 | 2105 | 6067371 |