Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317926.1 | NP_001304855.1 | 22 | 3154 | ||
NM_013355.4 | NP_037487.2 | 22 | 3198 | |||
XM_005251946.3 | XP_005252003.1 | 22 | 3753 | |||
XM_006717080.2 | XP_006717143.1 | 22 | 3267 | |||
XM_017014649.1 | XP_016870138.1 | 22 | 3225 | |||
XM_017014650.1 | XP_016870139.1 | 22 | 3407 | |||
GenBank mRNA | BC062558.1 | - | 22 | 2801 | 6649806 |