Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286756.1 | NP_001273685.1 | 20 | 1523 | ||
NM_013367.2 | NP_037499.2 | 20 | 1520 | |||
XM_005248159.2 | XP_005248216.1 | 7 | 417 | |||
XM_011513838.1 | XP_011512140.1 | 20 | 1400 | |||
XM_011513839.1 | XP_011512141.1 | 20 | 1444 | |||
XM_017008138.1 | XP_016863627.1 | 20 | 1449 | |||
GenBank mRNA | AF191338.1 | - | 19 | 1391 | ||
AK023597.1 | - | 17 | 1134 | |||
AK292336.1 | - | 20 | 1440 | |||
AL353932.1 | - | 3 | 93 | |||
BC059383.1 | - | 20 | 1493 | 30349524 |