Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191322.1 | NP_001178251.1 | 3 | 1228 | ||
NM_001191323.1 | NP_001178252.1 | 3 | 1315 | |||
NM_013372.6 | NP_037504.1 | 2 | 1438 | |||
XM_005254301.2 | XP_005254358.1 | 2 | 1502 | |||
XM_017022077.1 | XP_016877566.1 | 2 | 1484 | |||
GenBank mRNA | AF110137.2 | - | 2 | 1341 | ||
AF154054.1 | - | 2 | 1407 | |||
AK095741.1 | - | 2 | 1282 | |||
AK095890.1 | - | 3 | 1315 | |||
AK095972.1 | - | 3 | 1228 | |||
AL359060.1 | - | 1 | 33 | |||
AY232290.1 | - | 2 | 1414 |