Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005030.5 | NP_005021.2 | 5 | 1140 | ||
GenBank mRNA | AB209179.1 | - | 4 | 2999 | ||
AK303263.1 | - | 6 | 756 | |||
AK308276.1 | - | 6 | 997 | |||
AK313227.1 | - | 5 | 1064 | |||
BC002369.2 | - | 5 | 1032 | 2822226 | ||
BC003002.1 | - | 5 | 1010 | 2823623 | ||
BC014846.1 | - | 5 | 985 | 3854860 | ||
L19559.1 | - | 5 | 1042 | |||
U01038.1 | - | 5 | 1042 | |||
X73458.1 | - | 5 | 1056 | |||
X75932.1 | - | 5 | 1042 |