Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318024.1 | NP_001304953.1 | 12 | 1261 | ||
NM_013385.4 | NP_037517.1 | 12 | 1242 | |||
XM_011530147.2 | XP_011528449.1 | 12 | 1129 | |||
XM_011530148.1 | XP_011528450.1 | 13 | 1206 | |||
GenBank mRNA | AF075458.1 | - | 12 | 1171 | ||
AF125349.1 | - | 12 | 1242 | |||
AK002145.1 | - | 6 | 558 | |||
AK024428.1 | - | 11 | 2501 | |||
AK304453.1 | - | 12 | 1162 | |||
AK315052.1 | - | 12 | 1078 | |||
BC041161.1 | - | 12 | 1100 | 5745783 | ||
CR456551.1 | - | 12 | 1091 |