Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101648.1 | NP_001095118.1 | 2 | 130 | ||
NM_001300967.1 | NP_001287896.1 | 2 | 130 | |||
NM_013389.2 | NP_037521.2 | 2 | 130 | |||
XM_011515326.2 | XP_011513628.1 | 2 | 176 | |||
XM_011515327.2 | XP_011513629.1 | 2 | 465 | |||
GenBank mRNA | AF192522.1 | - | 2 | 130 | ||
AF192523.1 | - | 2 | 130 | |||
AY437865.1 | - | 2 | 74 | |||
AY515256.1 | - | 2 | 74 | |||
BC117178.1 | - | 2 | 128 | 40125729 | ||
BC143756.1 | - | 2 | 128 | 9052270 | ||
FJ481111.1 | - | 2 | 74 |