Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199125.1 | NP_001186054.1 | 5 | 436 | ||
NM_001316354.1 | NP_001303283.1 | 5 | 570 | |||
NM_001316355.1 | NP_001303284.1 | 5 | 483 | |||
NM_001316356.1 | NP_001303285.1 | 4 | 353 | |||
NM_001316357.1 | NP_001303286.1 | 4 | 433 | |||
NM_001316358.1 | NP_001303287.1 | 3 | 303 | |||
NM_003653.3 | NP_003644.2 | 5 | 483 | |||
XM_005256837.4 | XP_005256894.1 | 5 | 502 | |||
XM_005256840.3 | XP_005256897.1 | 5 | 547 | |||
XM_005256842.4 | XP_005256899.1 | 4 | 415 | |||
XM_017025246.1 | XP_016880735.1 | 4 | 341 | |||
GenBank mRNA | AF031647.1 | - | 5 | 388 | ||
AF098109.1 | - | 5 | 408 | |||
AK297706.1 | - | 5 | 458 | |||
AK302304.1 | - | 5 | 436 | |||
AK312476.1 | - | 5 | 424 | |||
AK316400.1 | - | 5 | 515 | |||
BC001891.1 | - | 5 | 457 | 3537198 | ||
BG030374.1 | - | 186 | ||||
BI520077.1 | - | 5 | 374 | |||
BP260214.1 | - | 5 | 483 | |||
BU154357.1 | - | 4 | 394 | |||
BU171304.1 | - | 4 | 267 | |||
DB493356.1 | - | 5 | 482 |