Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321357.1 | NP_001308286.1 | 17, 18 | 2277 | ||
NM_001321358.1 | NP_001308287.1 | 17, 18 | 1674 | |||
NM_001321359.1 | NP_001308288.1 | 17, 18 | 1669 | |||
NM_001321361.1 | NP_001308290.1 | 17, 18 | 1671 | |||
NM_001321362.1 | NP_001308291.1 | 16, 17 | 1647 | |||
NM_001321363.1 | NP_001308292.1 | 16, 17 | 1554 | |||
NM_013392.3 | NP_037524.1 | 16, 17 | 1645 | |||
GenBank mRNA | AF113249.1 | - | 16, 17 | 1557 | ||
AK001946.1 | - | 16, 17 | 1535 | |||
AK027538.1 | - | 16, 17 | 1553 | |||
AK092860.1 | - | 9, 10 | 1955 | |||
AK122664.1 | - | 17, 18 | 2277 | |||
AK130619.1 | - | 9, 10 | 689 | |||
AK223136.1 | - | 16, 17 | 1541 | |||
AK301345.1 | - | 17, 18 | 1488 | |||
AL136682.1 | - | 16, 17 | 1532 | |||
BC001221.1 | - | 16, 17 | 1559 | 3534543 | ||
BQ225378.1 | - | 2, 3 | 94 |