Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134493.1 | NP_001127965.1 | 3 | 718 | ||
GenBank mRNA | AJ420506.1 | - | 3 | 1390 | ||
BC015975.1 | - | 3 | 501 | |||
BC018624.1 | - | 3 | 446 | |||
BC022274.1 | - | 3 | 440 | |||
BC107435.1 | - | 3 | 457 | |||
L34839.1 | - | 3 | 440 |