Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003654.5 | NP_003645.1 | 4 | 1595 | ||
XM_006718356.3 | XP_006718419.1 | 4 | 1560 | |||
XM_017018459.1 | XP_016873948.1 | 4 | 1601 | |||
GenBank mRNA | AB003791.1 | - | 4 | 1256 | ||
AF090137.1 | - | 1 | 894 | |||
BC022567.1 | - | 4 | 1561 | 4814499 | ||
BC028235.1 | - | 4 | 1229 | 5245338 | ||
U65637.1 | - | 2 | 1014 |