Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039877.1 | NP_001034966.1 | 12 | 1633 | ||
NM_013403.2 | NP_037535.2 | 12 | 1612 | |||
XM_006723171.2 | XP_006723234.1 | 13 | 1690 | |||
XM_006723172.2 | XP_006723235.1 | 13 | 1669 | |||
XM_006723174.2 | XP_006723237.1 | 12 | 1440 | |||
XM_011526878.1 | XP_011525180.1 | 13 | 1814 | |||
XM_011526879.1 | XP_011525181.1 | 6 | 777 | |||
XM_017026716.1 | XP_016882205.1 | 13 | 1419 | |||
XM_017026717.1 | XP_016882206.1 | 12 | 1395 | |||
XM_017026718.1 | XP_016882207.1 | 8 | 1184 | |||
XM_017026719.1 | XP_016882208.1 | 12 | 1769 | |||
GenBank mRNA | AB209003.1 | - | 12 | 1585 | ||
AF212940.1 | - | 12 | 1579 | |||
AK056294.1 | - | 6 | 634 | |||
AK092913.1 | - | 3 | 948 | |||
AK225493.1 | - | 6 | 608 | |||
AK297637.1 | - | 13 | 1591 | |||
AK308813.1 | - | 7 | 847 | |||
BC004910.2 | - | 4 | 367 | |||
BC034604.1 | - | 12 | 1589 | 4829172 |