Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013438.4 | NP_038466.2 | 3 | 949 | ||
NM_053067.2 | NP_444295.1 | 3 | 949 | |||
XM_005251948.2 | XP_005252005.1 | 3 | 679 | |||
GenBank mRNA | AB035275.1 | - | 3 | 545 | ||
AB209468.1 | - | 2 | 93 | |||
AF176069.3 | - | 3 | 375 | |||
AF293384.1 | - | 3 | 375 | |||
AK027556.1 | - | 3 | 648 | |||
AK027576.1 | - | 3 | 639 | |||
AK074535.1 | - | 3 | 901 | |||
BC010066.2 | - | 3 | 629 | 4110398 | ||
BC039294.1 | - | 3 | 640 | 5263680 |