Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013438.4 | NP_038466.2 | 4 | 1110 | ||
NM_053067.2 | NP_444295.1 | 4 | 1110 | |||
XM_005251948.2 | XP_005252005.1 | 4 | 840 | |||
GenBank mRNA | AB035275.1 | - | 4 | 706 | ||
AB209468.1 | - | 3 | 254 | |||
AF176069.3 | - | 4 | 536 | |||
AF293384.1 | - | 4 | 536 | |||
AK027556.1 | - | 4 | 809 | |||
AK027576.1 | - | 4 | 800 | |||
AK074535.1 | - | 4 | 1062 | |||
AL136643.1 | - | 4 | 814 | |||
BC010066.2 | - | 4 | 790 | 4110398 | ||
BC039294.1 | - | 4 | 801 | 5263680 |